siRNA Details
| Gene Symbol: GS1-279B7.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038424.1 | 3 | 377 | |||
| GenBank mRNA | AA401571.1 | - | 3 | 377 | ||
| AF508905.1 | - | 3 | 345 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GS1-279B7.1| Gene Symbol: GS1-279B7.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038424.1 | 3 | 377 | |||
| GenBank mRNA | AA401571.1 | - | 3 | 377 | ||
| AF508905.1 | - | 3 | 345 | |||