siRNA Details
| Gene Symbol: LOC338694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104161.1 | 3 | 875 | |||
| GenBank mRNA | BC043531.1 | - | 3 | 875 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000562772.1 | - | 3 | 877 | Chr. 11 - 69147228 - 69171564 [-] on Build GRCh38 | |

LOC338694
Genomic Map