siRNA Details
| Gene Symbol: LOC101927087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110838.1 | 9 | 779 | |||
| GenBank mRNA | AK057455.1 | - | 9 | 779 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000563724.1 | - | 9 | 779 | Chr. 4 - 124499942 - 124558434 [-] on Build GRCh38 | |

LOC101927087
Genomic Map