siRNA Details
Gene Symbol: LOC101927087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110838.1 | 9 | 1111 | |||
GenBank mRNA | AK057455.1 | - | 9 | 1111 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000563724.1 | - | 9 | 1111 | Chr. 4 - 124499942 - 124558434 [-] on Build GRCh38 |
