siRNA Details
| Gene Symbol: LMO7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120410.1 | 3 | 295 | |||
| NR_120411.1 | 2 | 251 | ||||
| GenBank mRNA | AI816891.1 | - | 3 | 295 | ||
| AW188034.1 | - | 2 | 251 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LMO7-AS1| Gene Symbol: LMO7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120410.1 | 3 | 295 | |||
| NR_120411.1 | 2 | 251 | ||||
| GenBank mRNA | AI816891.1 | - | 3 | 295 | ||
| AW188034.1 | - | 2 | 251 | |||