siRNA Details
Gene Symbol: LOC100505853 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134646.1 | 2 | 599 | |||
GenBank mRNA | BC037384.1 | - | 2 | 581 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000564156.1 | - | 2 | 581 | Chr. 18 - 75696079 - 75712385 [-] on Build GRCh38 |
