siRNA Details
| Gene Symbol: LOC101927132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110650.1 | 4 | 1763 | |||
| GenBank mRNA | AK091370.1 | - | 4 | 1765 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000569726.1 | - | 4 | 1765 | Chr. 16 - 47858579 - 47908431 [+] on Build GRCh38 | |

LOC101927132
Genomic Map