siRNA Details
| Gene Symbol: LOC101929288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135005.1 | 2 | 401 | |||
| GenBank mRNA | BC039538.1 | - | 2 | 401 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000569178.1 | - | 2 | 401 | Chr. 20 - 7302056 - 7307432 [-] on Build GRCh38 | |

LOC101929288
Genomic Map