siRNA Details
Gene Symbol: LOC101929288 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135005.1 | 2 | 246 | |||
GenBank mRNA | BC039538.1 | - | 2 | 246 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000569178.1 | - | 2 | 246 | Chr. 20 - 7302056 - 7307432 [-] on Build GRCh38 |
