siRNA Details
| Gene Symbol: LOC105372440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134883.1 | 2 | 170 | |||
| NR_134884.1 | 2 | 170 | ||||
| GenBank mRNA | BX104195.1 | - | 2 | 77 | ||
| GQ868703.1 | - | 2 | 170 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105372440| Gene Symbol: LOC105372440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134883.1 | 2 | 170 | |||
| NR_134884.1 | 2 | 170 | ||||
| GenBank mRNA | BX104195.1 | - | 2 | 77 | ||
| GQ868703.1 | - | 2 | 170 | |||