siRNA Details
| Gene Symbol: PYCARD-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102400.1 | 2 | 855 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000561916.2 | - | 2 | 855 | Chr. 16 - 31201885 - 31203452 [+] on Build GRCh38 | |

PYCARD-AS1
Genomic Map