siRNA Details
| Gene Symbol: LINC00559 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047489.1 | 2 | 2732 | |||
| GenBank mRNA | BC043442.1 | - | 2 | 2735 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000569476.2 | - | 2 | 2734 | Chr. 13 - 90060252 - 90119719 [-] on Build GRCh38 | |

LINC00559
Genomic Map