siRNA Details
Gene Symbol: LOC202025 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL713660.1 | - | 2 | 3433 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000561486.1 | - | 2 | 3429 | Chr. 4 - 10068089 - 10073019 [-] on Build GRCh38 |
