siRNA Details
| Gene Symbol: LOC101929164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110746.1 | 4 | 975 | |||
| GenBank mRNA | AL133086.1 | - | 4 | 975 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000565584.1 | - | 4 | 975 | Chr. 19 - 23015075 - 23023697 [-] on Build GRCh38 | |

LOC101929164
Genomic Map