siRNA Details
| Gene Symbol: TTC23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC034816.1 | - | 2 | 2237 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000564527.1 | - | 2 | 2239 | Chr. 15 - 99139317 - 99145370 [+] on Build GRCh38 | |

TTC23
Genomic Map