siRNA Details
| Gene Symbol: LOC101927009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135195.1 | 3 | 3448 | |||
| GenBank mRNA | CR749525.1 | - | 3 | 3451 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000564305.1 | - | 3 | 3448 | Chr. 16 - 9355588 - 9408093 [+] on Build GRCh38 | |

LOC101927009
Genomic Map