siRNA Details
| Gene Symbol: LINC00560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC043278.2 | - | 2 | 1440 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000564593.1 | - | 2 | 1440 | Chr. 13 - 88626853 - 88630397 [+] on Build GRCh38 | |

LINC00560
Genomic Map