siRNA Details
| Gene Symbol: LOC101928682 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057367.1 | - | 2 | 1462 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000569872.1 | - | 2 | 1462 | Chr. 16 - 87267258 - 87292438 [-] on Build GRCh38 | |

LOC101928682
Genomic Map