siRNA Details
Gene Symbol: LINC01477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110791.1 | 1 | 463 | |||
GenBank mRNA | BC034625.1 | - | 1 | 463 | ||
BC045816.1 | - | 1 | 463 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000566101.1 | - | 1 | 463 | Chr. 18 - 40066286 - 40099233 [+] on Build GRCh38 |
