siRNA Details
Gene Symbol: LOC100508631 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134669.1 | 2 | 223 | |||
GenBank mRNA | BC042993.1 | - | 2 | 192 | ||
BG621125.1 | - | 2 | 224 | |||
BU568933.1 | - | 172 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000562049.1 | - | 2 | 186 | Chr. 4 - 37001816 - 37020708 [+] on Build GRCh38 |
