siRNA Details
| Gene Symbol: LOC100505817 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038340.1 | 3 | 1037 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000563172.1 | - | 3 | 1037 | Chr. 18 - 73324941 - 73349878 [+] on Build GRCh38 | |

LOC100505817
Genomic Map