siRNA Details
| Gene Symbol: LOC101927876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125946.1 | 3 | 2171 | |||
| GenBank mRNA | BC035225.1 | - | 3 | 2171 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000569711.1 | - | 3 | 2171 | Chr. 1 - 18065657 - 18074412 [+] on Build GRCh38 | |

LOC101927876
Genomic Map