siRNA Details
| Gene Symbol: LOC101927580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110917.1 | 4 | 1517 | |||
| GenBank mRNA | CR936694.1 | - | 4 | 1515 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000568279.2 | - | 4 | 1517 | Chr. 16 - 59855353 - 60053971 [+] on Build GRCh38 | |

LOC101927580
Genomic Map