siRNA Details
| Gene Symbol: LOC101927348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110915.1 | 3 | 482 | |||
| GenBank mRNA | BC039386.1 | - | 3 | 482 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927348| Gene Symbol: LOC101927348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110915.1 | 3 | 482 | |||
| GenBank mRNA | BC039386.1 | - | 3 | 482 | ||