siRNA Details
| Gene Symbol: MMP25-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_123723.1 | 5 | 1289 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000573130.2 | - | 2355 | Chr. 16 - 3053367 - 3056232 [-] on Build GRCh38 | ||

MMP25-AS1
Genomic Map