siRNA Details
| Gene Symbol: LOC101927727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135633.1 | 2 | 354 | |||
| GenBank mRNA | BG617385.1 | - | 2 | 110 | ||
| BQ934941.1 | - | 2 | 354 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000576252.1 | - | 2 | 354 | Chr. 17 - 909632 - 911212 [+] on Build GRCh38 | |

LOC101927727
Genomic Map