siRNA Details
Gene Symbol: LOC105371592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136412.1 | 2 | 401 | |||
GenBank mRNA | BU581060.1 | - | 2 | 401 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000572876.1 | - | 2 | 401 | Chr. 17 - 2712309 - 2712833 [+] on Build GRCh38 |
