siRNA Details
| Gene Symbol: LOC105371592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136412.1 | 2 | 115 | |||
| GenBank mRNA | BU581060.1 | - | 2 | 115 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000572876.1 | - | 2 | 115 | Chr. 17 - 2712309 - 2712833 [+] on Build GRCh38 | |

LOC105371592
Genomic Map