siRNA Details
| Gene Symbol: LINC00514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033861.1 | 2 | 742 | |||
| GenBank mRNA | AK098017.1 | - | 2 | 742 | ||
| AK128266.1 | - | 1 | 136 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00514| Gene Symbol: LINC00514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033861.1 | 2 | 742 | |||
| GenBank mRNA | AK098017.1 | - | 2 | 742 | ||
| AK128266.1 | - | 1 | 136 | |||