siRNA Details
Gene Symbol: LOC101928738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110851.1 | 1 | 113 | |||
GenBank mRNA | BC044939.1 | - | 1 | 113 | ||
BM850955.1 | - | 1 | 96 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110851.1 | 1 | 113 | |||
GenBank mRNA | BC044939.1 | - | 1 | 113 | ||
BM850955.1 | - | 1 | 96 |