siRNA Details
Gene Symbol: LOC101928738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110851.1 | 2 | 966 | |||
GenBank mRNA | BC044939.1 | - | 2 | 966 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000574526.1 | - | 2 | 966 | Chr. 17 - 79919357 - 79924010 [+] on Build GRCh38 |
