siRNA Details
| Gene Symbol: LOC101928738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110851.1 | 2 | 1171 | |||
| GenBank mRNA | BC044939.1 | - | 2 | 1171 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000574526.1 | - | 2 | 1171 | Chr. 17 - 79919357 - 79924010 [+] on Build GRCh38 | |

LOC101928738
Genomic Map