siRNA Details
| Gene Symbol: LINC01569 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_039999.1 | 3 | 1176 | |||
| GenBank mRNA | AK094828.1 | - | 3 | 1176 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000573042.1 | - | 3 | 1176 | Chr. 16 - 4245825 - 4253789 [-] on Build GRCh38 | |

LINC01569
Genomic Map