siRNA Details
| Gene Symbol: GLIS2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110901.1 | 4 | 267 | |||
| GenBank mRNA | BX104227.1 | - | 4 | 267 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000576080.1 | - | 4 | 267 | Chr. 16 - 4324667 - 4328340 [-] on Build GRCh38 | |

GLIS2-AS1
Genomic Map