siRNA Details
| Gene Symbol: LOC101927131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110907.1 | 3 | 1928 | |||
| GenBank mRNA | BC035158.1 | - | 3 | 1931 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000572950.1 | - | 3 | 1928 | Chr. 16 - 11465260 - 11473174 [+] on Build GRCh38 | |

LOC101927131
Genomic Map