siRNA Details
Gene Symbol: RBFOX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126788.1 | - | 2 | 3435 | ||
BC140939.1 | - | 1 | 2492 | 9056751 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000571452.1 | - | 2 | 3435 | Chr. 17 - 79598032 - 79603921 [-] on Build GRCh38 |
