siRNA Details
Gene Symbol: THCAT158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110880.1 | 3 | 618 | |||
NR_110881.1 | 3 | 482 | ||||
GenBank mRNA | AA430297.1 | - | 3 | 482 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000576345.2 | - | 2 | 420 | Chr. 17 - 47303466 - 47305685 [-] on Build GRCh38 |
