siRNA Details
| Gene Symbol: TAPT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027696.1 | 2 | 556 | |||
| NR_027697.1 | 2 | 556 | ||||
| GenBank mRNA | BF732723.1 | - | 2 | 230 | ||
| DA640966.1 | - | 2 | 179 | |||
| DR004294.1 | - | 2 | 540 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAPT1-AS1| Gene Symbol: TAPT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027696.1 | 2 | 556 | |||
| NR_027697.1 | 2 | 556 | ||||
| GenBank mRNA | BF732723.1 | - | 2 | 230 | ||
| DA640966.1 | - | 2 | 179 | |||
| DR004294.1 | - | 2 | 540 | |||