siRNA Details
Gene Symbol: LOC100507002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110801.1 | 3 | 539 | |||
GenBank mRNA | DA745846.1 | - | 3 | 539 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100507002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110801.1 | 3 | 539 | |||
GenBank mRNA | DA745846.1 | - | 3 | 539 |