siRNA Details
Gene Symbol: LOC339298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040034.1 | 1 | 493 | |||
GenBank mRNA | BC043002.2 | - | 1 | 483 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000584677.1 | - | 1 | 493 | Chr. 18 - 76122998 - 76145255 [+] on Build GRCh38 |
