siRNA Details
| Gene Symbol: PPP4R1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110773.1 | 3 | 437 | |||
| NR_110774.1 | 3 | 301 | ||||
| GenBank mRNA | BX108181.1 | - | 3 | 301 | ||
| HY048010.1 | - | 3 | 438 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000582435.1 | - | 3 | 301 | Chr. 18 - 9615264 - 9619363 [+] on Build GRCh38 | |

PPP4R1-AS1
Genomic Map