siRNA Details
Gene Symbol: LOC285766 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126020.1 | 5, 6 | 1503 | |||
GenBank mRNA | AK097546.1 | - | 5, 6 | 1503 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000580119.1 | - | 5, 6 | 1503 | Chr. 6 - 181466 - 205484 [-] on Build GRCh38 |
