siRNA Details
| Gene Symbol: LOC285766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126020.1 | 7 | 1808 | |||
| GenBank mRNA | AK097546.1 | - | 7 | 1808 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000580119.1 | - | 7 | 1808 | Chr. 6 - 181466 - 205484 [-] on Build GRCh38 | |

LOC285766
Genomic Map