siRNA Details
| Gene Symbol: LINC00908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015417.1 | 3 | 1364 | |||
| GenBank mRNA | BC121150.1 | - | 1 | 821 | ||
| BC121151.1 | - | 1 | 821 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000578613.5 | - | 3 | 1364 | Chr. 18 - 76528655 - 76559826 [+] on Build GRCh38 | |

LINC00908
Genomic Map