siRNA Details
| Gene Symbol: LINC00672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038847.1 | 2 | 1589 | |||
| GenBank mRNA | AK090935.1 | - | 2 | 1185 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00672| Gene Symbol: LINC00672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038847.1 | 2 | 1589 | |||
| GenBank mRNA | AK090935.1 | - | 2 | 1185 | ||