siRNA Details
Gene Symbol: LOC100505797 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134647.1 | 3 | 319 | |||
NR_134648.1 | 2 | 145 | ||||
GenBank mRNA | BX114234.1 | - | 3 | 319 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100505797 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134647.1 | 3 | 319 | |||
NR_134648.1 | 2 | 145 | ||||
GenBank mRNA | BX114234.1 | - | 3 | 319 |