siRNA Details
| Gene Symbol: LOC146795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135222.1 | 2 | 1407 | |||
| GenBank mRNA | AK057377.1 | - | 2 | 1279 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000577573.1 | - | 2 | 1278 | Chr. 17 - 72342940 - 72355136 [+] on Build GRCh38 | |

LOC146795
Genomic Map