siRNA Details
Gene Symbol: LOC146795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135222.1 | 3, 4 | 1735 | |||
GenBank mRNA | AK057377.1 | - | 3, 4 | 1607 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000577573.1 | - | 3, 4 | 1606 | Chr. 17 - 72342940 - 72355136 [+] on Build GRCh38 |
