siRNA Details
| Gene Symbol: LOC104968399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130143.1 | 3 | 785 | |||
| NR_130144.1 | 3 | 467 | ||||
| NR_130145.1 | 4 | 592 | ||||
| GenBank mRNA | BG201476.1 | - | 2 | 232 | ||
| BG218162.1 | - | 2 | 241 | |||
| BG721448.1 | - | 3 | 469 | |||

LOC104968399
Genomic Map