siRNA Details
Gene Symbol: LOC104968399 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130143.1 | 3 | 733 | |||
NR_130144.1 | 3 | 415 | ||||
NR_130145.1 | 4 | 540 | ||||
GenBank mRNA | BG218162.1 | - | 2 | 189 | ||
BG721448.1 | - | 3 | 417 | |||
BG773124.1 | - | 3 | 731 |
